Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC Universal Lintfree UN810 Boom

Catalog No. 17111094 Shop All Brady Corporation Products
Click to view available options
:

Universal absorbent booms are ideal for containment both indoors and out.

  • Universal Plus booms can be used on land and indoors to absorb all water-based, oil-based and chemical fluids
  • Universal absorbency - picks up water-based, petroleum-based and non-aggressive, chemical fluids
  • Booms are lint-free and easy to handle
  • Included clips allow for connection to extend overall length

Specifications

Height (English)

8 in.

Height (Metric)

20.3 cm

Type

Universal Absorbent Boom

Length (English)

10 ft.

Diameter (English)

8 in.

Color

Green

Length (Metric)

3.04 m

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.