Learn More
NEOGEN Brain Heart Infusion Agar, 5 kg
SDP
Supplier:  NEOGEN 700003194
Encompass_Preferred
Brain Heart Infusion Agar is used for the cultivation of a wide variety of fastidious organisms and is not intended for use in the diagnosis of disease or other conditions in humans.A general purpose nutritious agar base. This medium was first used for the isolation of dental pathogens. With the addition of 7% defibrinated blood the medium will support the growth of a wide range of fastidious organisms, the phosphate buffer will help neutralize the acids produced from the utilization of glucose and thus maintain viability. The medium is not recommended for the determination of hemolytic reactions because of the glucose content.
 For more information visit https://www.neogen.com/solutions/Microbiology/brain-heart-infusion-agar
Catalog No. 50-201-5388
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.