Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brass foil, alloy 260, 0.81mm (0.032 in.) thick, Thermo Scientific™

Catalog No. p-7059688
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Brass foil, Alloy 260
Name Note 0.81mm (0.032 in.) thick
MDL Number MFCD00171255
Quantity 150 x 150 mm
Odor Odorless
Form Foil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.