Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is desig
SDP
Supplier:  Bellco Glass, Inc. 254100250
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is desig
Catalog No. 50-194-5519