Learn More
Chemglass Life Sciences Flask, Shake, 2000mL, Deep Baffles, Plain Top, Approx OD x Height (mm): 155 x 270
SDP
Supplier:  Chemglass Life Sciences CLS204207
Encompass_Preferred
Flask, Shake, 2000mL, Deep Baffles, Plain Top, Approx OD x Height (mm): 155 x 270. Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
Catalog No. 50-121-4771
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.