Learn More
Selleck Chemical LLC Bromodeoxyuridine (BrdU) 200mg 59-14-3
Supplier:  Selleck Chemical LLC S7918200MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Bromodeoxyuridine (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA, and used in the detection of proliferating cells.
Catalog No. 50-136-5414
missing translation for 'QuantityLeft'