Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromopentacarbonylmanganese(I), 98%, Thermo Scientific Chemicals

Catalog No. p-7050709
Click to view available options
Quantity:
1 g
5 g

Bromopentacarbonylmanganese(I) is used in the formation of (eta6-arene)tricarbonylmanganese(I) by reacting with arene (arene= hexamethyl benzene, 1,2,4,5-tetramethyl benzene, mesitylene, p-xylene and toluene) in the presence silver salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14516-54-2
Molecular Formula C5BrMnO5-
Molecular Weight (g/mol) 274.892
MDL Number MFCD00049806
InChI Key LOCZKKJCQVRJPQ-UHFFFAOYSA-M
Synonym bromopentacarbonylmanganese i
PubChem CID 10978692
IUPAC Name carbon monoxide;manganese;bromide
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mn].[Br-]

Specifications

Color Orange
Linear Formula Mn(CO)5Br
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 274.89
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Bromopentacarbonylmanganese(I)
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 238-522-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.