Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromopentafluorobenzene 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | -31°C |
Color | Yellow |
Boiling Point | 137°C |
Quantity | 5 g |
Formula Weight | 246.96 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Bromopentafluorobenzene |
Safety and Handling
EINECSNumber | (3)-4285 |
RTECSNumber | CY9040552 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction