Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bronze gauze, alloy 220, 80 mesh woven from 0.14mm (0.0055in) in dia wire, Thermo Scientific™
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Chemical Name or Material | Bronze gauze, Alloy 220 |
Name Note | 80 mesh woven from 0.14mm (0.0055 in.) dia. wire |
MDL Number | MFCD00198187 |
Quantity | 150 x 150 mm |
Mesh Size | 80 Mesh |
Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction