Learn More
Carolina™ Bruno Lo Carbon Composite Toe
Men's 6 Inch ESD Waterproof Carbon Composite Broad Toe Work Boot
Supplier: Carolina™ CA55221002E
This item is not returnable.
View return policy
Description
- Brown
- Gaucho Crazy Horse Leather
- Composite Safety Toe
- EH
- 8 Inch
- 400G Thinsulate
Specifications
Brown | |
10 | |
Composite |
Leather | |
Rubber | |
2E |