Learn More
Bellco Glass, Inc. Brush-Less Digital Roller Drum 230V
SDP
Supplier:  Bellco Glass, Inc. 773631230
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Bellco Brushless Digital Roller Drum Drive is an all purpose roller drum for blood, tissue culture, cells and virus applications. The standard speed ranges are 0.5 to 15 and 2 to 60 RPM with other speed ranges available as custom items. Four drums ar
Catalog No. 50-194-6093
missing translation for 'QuantityLeft'