Learn More
Electron Microscopy Sciences Aurion BSA Gold Tracer, UltraSmall 10 ML
Supplier: Electron Microscopy Sciences 25480
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones, cationic tracers bind to polyanionic moieties (membranes, acidic proteins). "Neutral" BSA coated gold tracers are useful for detecting open connections and tissue damage. Under proper conditions and with suitable microscopical techniques the tracers can also be used to follow cellular events in time.
Anionic Gold Tracers are prepared with BSA-c as particle conjugated protein. AURION Gold Tracers are shipped containing NaN3 as preservative. If they are intended for use in living organisms, the preservative has to be removed prior to use.
Particle size: UltraSmall
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.