Learn More
G-Biosciences Bsu DNA Polymerase, Large Fragment
SCI_ED
Bsu DNA Polymerase, Large fragment is used for second strand cDNA synthesis, strand displacement DNA synthesis, random primer labeling and dA-tailing.
Supplier:  G-Biosciences 7861999
 View more versions of this product
Catalog No. S03118
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Bsu DNA Polymerase Large fragment is from Bacillus subtilis and is modified to retain its 5' - 3' DNA polymerase activity with its 5' - 3' exonuclease domain removed.
  • It also lacks the 3' - 5' exonuclease activity.
Specifications
Specifications
Gel Electrophoresis
High
Standard
DNA Polymerase
1000
Liquid
Standard PCR, qPCR, Multiplex qPCR, RT-PCR, 1-step RT-PCR
Biotin, FAM, 6-FAM, SYBR Green, Unlabeled
Microbial Detection, Pathogen Detection, Virus Detection, Genetic Disease Research, Isothermal Amplification
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.