Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTZO 1, Tocris Bioscience™

Catalog No. p-4817326 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MIF binder

Macrophage migration inhibitory factor (MIF) binder (Kd = 68.6 nM). Activates antioxidant response element (ARE)-mediated gene expression. Suppresses oxidative stress-induced cardiomyocyte apoptosis in vitro. Cardioprotective.

Chemical Identifiers

CAS 99420-15-2
Molecular Formula C13H8N2OS
Molecular Weight (g/mol) 240.28
InChI Key GBAKVEWPYUIGHN-UHFFFAOYSA-N
Synonym btzo 1, 2-2-pyridyl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-4h-benzo e 1,3 thiazin-4-one, 2-pyridin-2-yl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-1,3-benzothiazin-4-one, btzo-1, btzo hplc, 2-2-pyridinyl-4h-1,3-benzothiazin-4-one
PubChem CID 9837640
IUPAC Name 2-pyridin-2-yl-1,3-benzothiazin-4-one
SMILES C1=CC=C2C(=C1)C(=O)N=C(S2)C3=CC=CC=N3

Specifications

Quantity 10 mg
Formula Weight 240.28
Percent Purity >99%
Chemical Name or Material BTZO 1
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.