Learn More
Medchemexpress LLC HY-N0877 5mg Medchemexpress, Bufalin CAS:465-21-4 Purity:>98%
Supplier: Medchemexpress LLC HYN08775MG
Medchemexpress, HY-N0877 5mg Bufalin CAS:465-21-4 Bufalin is an active component isolated from Chan Su, acts as a potent Na + /K + -ATPase inhibitor, binds to the subunit α1, α2 and α3, with K d of 42.5, 45 and 40 nM, respectively [1] [2] . Anti-cancer activity [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.