Learn More
NEOGEN Buffered Peptone Water (BPW), 500 g
SDP
Supplier:  NEOGEN 700002954
Encompass_Preferred
Buffered Peptone Water (BPW) is used for the non-selective pre-enrichment of Salmonella spp. from food and is not intended for use in the diagnosis of disease or other conditions in humans. Edel and Kamelmacher found that food preservation techniques involving heat, desiccation, preservatives, high osmotic pressure, or pH changes cause sublethal injury to Salmonella spp. Preenrichment in a non-selective medium allows for repair of cell damage and facilitates the recovery of Salmonella. Lactose Broth is frequently used for this purpose, but it may be detrimental to recovering Salmonellae. Buffered Peptone Water maintains a high pH over the pre-enrichment period and allows in repair of injured cells that may be sensitive to low pH. This is particularly important for vegetable specimens which have a low buffering capacity. Buffered Peptone Water is used in standard methods.  For more information visit https://www.neogen.com/solutions/Microbiology/buffered-peptone-water
Catalog No. 50-201-5162
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.