Learn More
BUV395 Biosimilar Anti-Human CD20 Rituximab297, BD Horizon™
Biosimilar Monoclonal Antibody
Supplier: BD Biosciences 570430
Description
CD20 Rituximab297 Monoclonal antibody specifically detects CD20 Rituximab297 in Human samples. It is validated for Flow cytometrySpecifications
CD20 Rituximab297 | |
Monoclonal | |
Brilliant Ultraviolet 395 | |
B1; Bp35; LEU-16; MS4A1 | |
Human CD20 | |
25 Tests | |
931 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
Rituximab297.rMAb (also known as Rituximab N297A Biosimilar.rMAb) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.