Learn More
BUV395 Mouse Anti-Human CD47, BD Horizon™
Mouse Monoclonal Antibody
$162.00 - $396.00
Specifications
Antigen | CD47 |
---|---|
Clone | CC2C6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow cytometrySpecifications
CD47 | |
Flow Cytometry | |
Brilliant Ultraviolet 395 | |
Mouse | |
Human | |
CD47; IAP; integrin associated protein; MER6; OA3; Rh-related antigen | |
Human CCRF-CEM | |
Affinity Chromatography |
CC2C6 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
961 | |
IgG1 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.