Learn More
BUV805 Mouse Anti-Biotin, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 758027
Description
Biotin Monoclonal antibody specifically detects Biotin. It is validated for Flow cytometrySpecifications
Biotin | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Biotin-conjugated Keyhole Limpet Hemocyanin (KLH) | |
50 μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
BK-1/39 | |
Brilliant Ultraviolet 805 | |
Mouse | |
Affinity Chromatography | |
RUO | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.