Learn More
BV650 Mouse Anti-Human TIGIT, BD Horizon™
Mouse Monoclonal Antibody
$178.00 - $442.00
Specifications
Antigen | TIGIT |
---|---|
Clone | TgMab-2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 650 |
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow cytometrySpecifications
TIGIT | |
Flow Cytometry | |
Brilliant Violet 650 | |
Mouse | |
Human | |
T-cell immunoreceptor with Ig and ITIM domains; VSIG9; VSTM3; WUCAM | |
Human TIGIT Transfected Cell Line | |
Affinity Chromatography |
TgMab-2 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
201633 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.