Learn More
BV786 Hamster Anti-Mouse CD11c (Integrin αX), BD Horizon™
Hamster Monoclonal Antibody
$288.00 - $480.00
Specifications
Antigen | CD11c (Integrin αX) |
---|---|
Clone | N418 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD11c (Integrin αX) Monoclonal antibody specifically detects CD11c (Integrin αX) in Mouse samples. It is validated for Flow cytometrySpecifications
CD11c (Integrin αX) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Dendritic Cells | |
Affinity Chromatography |
N418 | |
Flow Cytometry | |
Brilliant Violet 786 | |
Hamster | |
Mouse | |
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4 | |
IgG2 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.