Learn More
BV786 Rat Anti-Mouse CD138 (Syndecan-1), BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 569692
Description
CD138 (Syndecan-1) Monoclonal antibody specifically detects CD138 (Syndecan-1) in Mouse samples. It is validated for Flow cytometrySpecifications
CD138 (Syndecan-1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
281-2 | |
Brilliant Violet 786 | |
SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin | |
NAMRU mouse mammary gland epithelial cell line NMuMG | |
50 μg | |
20969 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.