Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BX 513 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective CCR1 receptor antagonist (Ki values are 0.04, > 10, > 10 and > 10 nM for CCR1, CCR5, CXCR2 and CXCR4 receptors respectively). Inhibits MIP-1α-induced intracellular calcium mobilization (IC50 = 2.5 μM). Also a full inverse agonist at US28, a HCMV-encoded chemokine receptor.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 481.46 |
Percent Purity | >99% |
Chemical Name or Material | BX 513 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction