Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BX 795, Tocris Bioscience™

Catalog No. p-4816784 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PDPK1 (PDK1) inhibitor

Inhibitor of 3-phosphoinositide-dependent kinase 1 (PDPK1). Inhibits Akt phosphorylation at Thr308 in PC3 cells; also inhibits anchorage-independent growth of PC3 and MDA-MB-468 cells. Exhibits activity at other kinases, including TANK-binding kinase 1 (TBK1), Aurora B and IκB kinase ε (IKKε). Also enhances lentiviral transduction of natural killer (NK) cells by around 3.8-fold.

Chemical Identifiers

CAS 702675-74-9
Molecular Formula C23H26IN7O2S
Molecular Weight (g/mol) 591.47
MDL Number MFCD12546134
InChI Key VAVXGGRQQJZYBL-UHFFFAOYSA-N
Synonym n-3-5-iodo-4-3-2-thienylcarbonyl amino propyl amino-2-pyrimidinyl amino phenyl-1-pyrrolidinecarboxamide, n-3-5-iodo-4-3-thiophene-2-carbonylamino propylamino pyrimidin-2-yl amino phenyl pyrrolidine-1-carboxamide, n-3-4-3-thiophene-2-carbonylamino propylamino-5-iodopyrimidin-2-ylamino phenyl pyrrolidine-1-carboxamide, n-3-5-iodo-4-3-thiophene-2-carboxamido propylamino pyrimidin-2-ylamino phenyl pyrrolidine-1-carboxamide, n-3-5-iodo-4-3-thiophen-2-ylcarbonyl amino propyl amino pyrimidin-2-yl amino phenyl pyrrolidine-1-carboxamide, n-3-5-iodo-4-3-thiophen-2-ylformamido propyl amino pyrimidin-2-yl amino phenyl pyrrolidine-1-carboxamide, bx7, d0rg0z, n-3-5-iodo-4-3-thiophene-2-carboxamido propyl amino pyrimidin-2-yl amino phenyl pyrrolidine-1-carboxamide, n-3-4-2-2-formamidothiophen-3-yl propanamido-5-iodopyrimidin-2-ylamino phenyl pyrrolidine-1-carboxamide
PubChem CID 10077147
IUPAC Name N-(3-{[5-iodo-4-({3-[(thiophen-2-yl)formamido]propyl}amino)pyrimidin-2-yl]amino}phenyl)pyrrolidine-1-carboxamide
SMILES IC1=CN=C(NC2=CC(NC(=O)N3CCCC3)=CC=C2)N=C1NCCCNC(=O)C1=CC=CS1

Specifications

Quantity 10 mg
Formula Weight 591.47
Percent Purity >98%
Chemical Name or Material BX 795
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.