Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C 75, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Synthetic inhibitor of fatty acid synthase (FASN); inhibits fatty acid synthesis in vitro and in vivo. Displays anorectic effects. Induces apoptosis in MCF-7 xenografts and exhibits anti-tumor activity.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 254.32 |
Percent Purity | >99% |
Chemical Name or Material | C 75 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction