Learn More
Medchemexpress LLC HY-145405 50mg , C12-200 CAS:1220890-25-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14540550MG
Encompass_Preferred
HY-145405 50mg C12-200 CAS: 1220890-25-4 C12-200 is a benchmark ionizable cationic lipidoid along with helper lipids. C12-200 is commonly used for mRNA delivery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0864
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.