Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CA(PEG)12 Carboxy-PEG-Amine Compound

Catalog No. p-7184462 Shop All Thermo Scientific Products
Click to view available options
Description:
Pierce CA(PEG)12
Quantity:
100 mg
1 g

CA(PEG)n: pegylated amino acids of the form carboxy-PEG-amine; with 4 to 24 polyethylene glycol units; for surface- and molecule-pegylation applications.

Thermo Scientific™ Pierce CA(PEG)n Compounds are pegylated amino acids of the form carboxy-PEG-amine, where n equals 4 to 24 polyethylene glycol units, and are useful for a variety of surface- and molecule-pegylation applications.

CA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with carboxy (-COOH) and amino (-NH2) termini. The unbranched, hydrophilic, discrete-length molecules have the form Carboxy-PEGn-Amine, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The carboxyl and primary amine of each compound provide specific targets for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.

Highlights:

  • Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Allows site-specific labeling or crosslinking involving primary amines or carboxyl groups on proteins or surfaces
  • Easy-to-follow instructions increase the likelihood of a successful outcome

Ordering Information:

Alternative names: Amino-dPEG-acid; Amino-PEG-carboxylic acid; Carboxy-PEO-amine

Compatible with:

Methyl-PEG-Amine Compounds, MA(PEG)n; Other Kinds of PEGylation Reagents

Specifications

Chemical Reactivity Aldehyde
Description Pierce CA(PEG)12
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Label Type Conjugating Groups, Polyethylene Glycol (PEG)
Reactive Moiety Amine, Carboxylic Acid, Primary Amine
Spacer 12 ethylene glycol units
Form Liquid
Quantity 100 mg
Solubility Water
Product Type Amine Compound

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.