Learn More
TARGETMOL CHEMICALS INC CABERGOLINE 10MG
Supplier:  TARGETMOL CHEMICALS INC T14853
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cabergoline (FCE-21336) is an ergot derived-dopamine D2-like receptor agonist. It has high affinity for D2 D3 and 5-HT2B receptors (Ki=0.7 1.5 and 1.2 respectively).Cabergoline permits rapid and effective hormonal and tumor control by normalizing prolactin-producing pituitary adenomas levels. purity: 99%
Catalog No. 50-257-7376
missing translation for 'QuantityLeft'