Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cabergoline, Tocris Bioscience™
GSA_VA

Catalog No. p-4783818 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D2-like agonist

Cabergoline is a selective D2-like dopamine receptor agonist (Ki values are 0.7, 1.5, 9.0 and 165 nM for D2, D3, D4 and D5 receptors respectively) that also displays high affinity for several serotonin receptor subtypes (Ki = 1.2 - 20.0 nM for 5-HT1A, 5-HT1D, 5-HT2A and 5-HT2B). Inhibits secretion of prolactin and growth hormone and reverses levodopa -induced dyskinesias in Parkinsonian monkeys.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 81409-90-7
Molecular Formula C26H37N5O2
Molecular Weight (g/mol) 451.615
InChI Key KORNTPPJEAJQIU-KJXAQDMKSA-N
Synonym cabergoline, dostinex, cabaser, cabergolina, cabergolinum, cabaseril, cabergolinum latin, cabergolina spanish, galastop, sogilen
PubChem CID 54746
ChEBI CHEBI:3286
IUPAC Name (6aR,9R,10aR)-N-[3-(dimethylamino)propyl]-N-(ethylcarbamoyl)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
SMILES CCNC(=O)N(CCCN(C)C)C(=O)C1CC2C(CC3=CNC4=CC=CC2=C34)N(C1)CC=C

Specifications

Quantity 10 mg
Formula Weight 451.6
Percent Purity >99%
Chemical Name or Material Cabergoline
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.