Learn More
3M™ Cable Tie
3M™ Standard Nylon Cable Ties are made with high-quality nylon 6/6 and designed to secure wire bundles and harness components quickly, without slipping. The curved tip allows for faster threading and installation, and they feature a self-locking head.
Supplier: 3M™ 7000133455
Description
- Secure wire bundles and harness components quickly, without slipping
- Curved tip allows for fasterthreading and installation
Specifications
Standard Cable Tie | |
UL Listed 62275 Type 21 or 21S and Plenum Rated AH-2 for use in air handling spaces | |
Electrical Construction and Maintenance, Mining and Metallurgy | |
Nylon 6/6 | |
147.32 mm |
CT6NT40-M | |
Black | |
500/Cs. | |
40 lb. | |
3.6 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.