Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Supplier: Cadence Science™ 7971
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
- Available with or without a reinforcing bar
- Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
22 G | |
Without reinforcing bar | |
Micro Emulsifying Needle w/o Reinforcing Bar | |
Non-sterile |
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar | |
2.87 in. | |
Glass |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.