Learn More
Cadmium powder, -200 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Quantity | 5 g |
UN Number | UN3179 |
IUPAC Name | cadmium(2+) ion |
Percent Purity | 99.99% |
Mesh Size | −200 Mesh |
Form | Solid |
Assay | (metals basis) |
Chemical Name or Material | Cadmium powder |
Safety and Handling
Hazard Category | H228-H330-H341-H350-H361fd-H372 |
Hazard Statement | GHS H Statement H330-H350-H372-H341-H361 Fatal if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. |
Precautionary Statement | P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S. |
EINECSNumber | 231-152-8 |
RTECSNumber | EU9800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.