Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Cage-Klenz 220 Acid-Based Detergent

Catalog No. p-200076289 Shop All STERIS Corporation Products
Click to view available options
Size:
Container Type:

Cage-Klenz 220 Acid-Based Detergent is a specially formulated glycolic acid detergent used for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging. pH (1%): 2.8, typical.

Cage-Klenz 220 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.

HOW CAGE-KLENZ 220 DETERGENT WORKS

Cage-Klenz 220 Detergent is a liquid phosphate-free, hydroxyacetic acid-based detergent.

WHY CAGE-KLENZ 220 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale, and organic soils in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties:

pH (1%): 2.8, typical

Specifications

Product Type

Acid Cleaner

Color-coded

No

Form

Clear Liquid

pH

2.8 (Diluted)

Scent

Unscented

Sterility

Non-sterile

Container Type

Pail

Size

5 gal.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.