Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Cage-Klenz 280 Acid-Based Detergent

Cage-Klenz 280 Detergent: Economical formula for hard water and urine scale removal in research animal caging. Phosphoric acid-based, materials-compatible, cost-saving, and environmentally friendly.

$253.00 - $1281.00

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.25 (Diluted)
Scent Unscented
View More Specs

Products 2
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
19-999-645
View Documents
STERIS™
1K2805
5 gal. Pail
Each for $253.00
missing translation for 'QuantityLeft'
Add to Cart
 
19-999-644
View Documents
STERIS™
1K2802
30 gal. Drum
Each for $1,281.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Cage-Klenz 280 Detergent is an economical detergent specially formulated to remove challenging hard water and urine scales found in research animal caging.

HOW CAGE-KLENZ 280 DETERGENT WORKS

Cage-Klenz 280 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.

WHY CAGE-KLENZ 280 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties: pH (1%): 2.25, typical

Specifications

Specifications

Detergent
Clear Liquid
Unscented
No
2.25 (Diluted)
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.