Learn More
Gasco 100 PPM Isobutylene in Air Calibration Gas, 34 L Steel Disposable Cylinder CGA 600 Connection
Supplier: Gasco 34LS248100
100 PPM Isobutylene, Balance Air, (20.9% Oxygen in Nitrogen) Calibration Gas contained in a 34 Liter Steel Disposable Cylinder.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.