Learn More
MilliporeSigma™ Calbiochem™ [6]-Gingerol, Zingiber officinale
Shop All MilliporeSigma ProductsSpecifications
Form
Oil
Content And Storage
Protect from light
Quantity
5 mg
Color
Pale Yellow
Description
95% by HPLC
Target Proteins
Activator Protein-1 (AP-1)
CAS
23513-14-6
Molecular Formula
C17H26O4
Formula Weight
294.4g/mol
Solubility
DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.