Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, 8-(4-Chlorophenylthio)-, Sodium Salt
Shop All MilliporeSigma Products
Click to view available options
:
10mg
Description
Cell Permeable
Specifications
Specifications
Content And Storage | Hygroscopic |
Quantity | 10 mg |
Form | Solid |
Color | White |
Description | 98% by HPLC |
Target Proteins | Protein kinase A and protein kinase G |
CAS | 93882-12-3 |
Molecular Formula | C16H14ClN5O6PS·Na |
Formula Weight | 493.8g/mol |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction