Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Adenosine 3′,5′-cyclic Monophosphorothioate, 8-Bromo-, Rp-Isomer, Sodium Salt
Shop All MilliporeSigma Products
Click to view available options
:
5 umol
Description
Cell Permeable
Specifications
Specifications
Content And Storage | Hygroscopic |
Quantity | 5 μmol |
Form | Lyophilized |
Description | 99% by HPLC |
Target Proteins | PKA 1 |
Molecular Formula | C10H10BrN5O5PS·Na |
Formula Weight | 446.1g/mol |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction