Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine A3 Receptor Agonist, 2-Cl-IB-MECA

Catalog No. 1191392MG Shop All MilliporeSigma Products
Click to view available options
:
2mg

A highly selective agonist of adenosine A3 receptor (Ki = 330 pM).

Packaged Under Inert Gas

Specifications

Quantity 2 mg
Form Solid
Content And Storage Protect from light
Color White
Description 98% by HPLC
CAS 163042-96-4
Molecular Formula C18H18ClIN6O4
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.