Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Adenosine A1 Receptor Agonist I, CPA
Shop All MilliporeSigma Products
Click to view available options
:
25mg
Description
A potent and selective agonist of adenosine A1 receptor (A1R) (Ki = 2.3 nM, 790 nM, 18.6μM, 43nM for human A1, A2A, A2B, and A3, respectively).
Specifications
Specifications
Form | Solid |
Content And Storage | Protect from light |
Quantity | 25 mg |
Color | White |
Description | 99% by HPLC |
Target Proteins | A1 |
CAS | 41552-82-3 |
Molecular Formula | C15H21N5O4 |
Formula Weight | 335.4g/mol |
Solubility | DMSO, 1N HCl, or EtOH |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction