Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine A1 Receptor Agonist II, CCPA

Catalog No. 11913610MG Shop All MilliporeSigma Products
Click to view available options
:
10mg

An adenosine analog that acts as a selective, high affinity agonist for Adenosine A1 receptor (A1R) (Ki = 800 pM

An adenosine analog that acts as a selective, high affinity agonist for Adenosine A1 receptor (A1R) (Ki = 800 pM; 2.3μM, 18.8μM, and 42nM for human A1, A2A, A2B, A3 respectively). Shown to be centrally active following systemic administration and exhibits anti-convulsive effects. Also reported to inhibit adenylate cyclase activity in rat fat cell membrane (IC50 = 33nM). Diminishes respiratory rhythm, reduces heart rate, and enhances the activity of KATP channels. Reduces norepinephrine release by about 50% in untreated and pertussis toxin treated hearts.

Specifications

Form Solid
Content And Storage Protect from light
Quantity 10 mg
Color White
Description 99% by HPLC
Target Proteins A1
Molecular Formula C15H20ClN5O4
Formula Weight 369.8g/mol
Solubility DMSO or H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.