Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine A2A Receptor Agonist I, CGS 21680, Hydrochloride

Catalog No. 1191375MG Shop All MilliporeSigma Products
Click to view available options
:
5mg

A xanthin derivative that acts as a potent agonist of adenosine A2A receptors (Ki =

A xanthin derivative that acts as a potent agonist of adenosine A2A receptors (Ki = 27nM). Exhibits reduced affinity for A2B receptors (Ki = 67nM), but offers greater selectivity over A1R (Ki = 290nM) and A3R (88.8μM). Exerts an anti-inflammatory effect during chronic inflammation and ameliorates the tissue damage associated with collagen-induced arthritis. Also shown to reduce CCR7 Protein Expression in THP1 macrophages. Shown to delay the progressive deterioration of motor performance and prevent a reduction in brain weight in murine model of Huntington's disease. Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Specifications

Content And Storage Protect from light
Quantity 5 mg
Form Solid
Color White
Formulation Supplied as a hydrochloride salt.
Description 98% by HPLC
Molecular Formula C23H29N7O6·HCl
Formula Weight 536g/mol
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.