Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine Receptor Agonist, NECA

Catalog No. 11914010MG Shop All MilliporeSigma Products
Click to view available options
:
10mg

A cell-permeable adenosine analog that acts a potent non-selective agonist of adenosine receptors (Ki = 14 nM, 20 nM

A cell-permeable adenosine analog that acts a potent non-selective agonist of adenosine receptors (Ki = 14 nM, 20 nM, 2.4μM and 6.2nM for A1, A2A, A2B, A3, respectively). Increases intracellular cAMP production (EC50 = 3.1μM in A2B expressing CHO cells). Shown to increase glucagon release in a dose-dependent manner and inhibit insulin release at low concentrations. Although at higher concentration some insulin release is observed. Also, displays a wide range adenosine-dependent effects, such as blocking platelet aggregation and inhibiting DNA synthesis. When administered at reperfusion, it is shown reduce infarction and block the formation of the mitochondrial permeability transition pore by activating p70S6 kinase.

Specifications

Content And Storage Protect from light
Form Solid
Quantity 10 mg
Color White
Description 99% by HPLC
Molecular Formula C12H16N6O4
Formula Weight 308.3g/mol
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.