Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Bupropion Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
:
50mg
Description
Deactivate with 5% Bleach
Specifications
Specifications
Form | Solid |
Content And Storage | Protect from light |
Quantity | 50 mg |
Color | White |
Description | 99% by HPLC |
Target Proteins | DAT/NET |
Molecular Formula | C13H18ClNO·HCl |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction