Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Citalopram Hydrobromide
Shop All MilliporeSigma Products
Click to view available options
:
25mg
Description
A potent uptake inhibitor highly selective for 5-HT transporter without effect on noradrenalin or dopamine uptake (IC50 = 1.8, 8800 and 41000nM for 5-HT, noradrenalin and dopamine, respectively). Clinically prescribed as an antidepressant drug. Widely used in research related to depressive disorders.
Specifications
Specifications
Quantity | 25 mg |
Form | Solid |
Content And Storage | Protect from light |
Color | White |
Description | 98% by HPLC |
Target Proteins | 5-HT Transporter |
Molecular Formula | C20H21FN2O·HBr |
Solubility | EtOH or H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction