Learn More
MilliporeSigma™ Calbiochem™ ET-18-OCH3
Shop All MilliporeSigma ProductsSpecifications
Form
Solid
Quantity
5 mg
Color
White
Description
95% by TLC
Target Proteins
PI-PLC
CAS
70641-51-9
Molecular Formula
C27H58NO6P
Formula Weight
523.7g/mol
Solubility
DMSO, CHCl3, EtOH, or H2O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.