Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GBR 12909 Dihydrochloride

Catalog No. 5057320001 Shop All MilliporeSigma Products
Click to view available options
:
10mg

A potent inhibitor of dopamine active transporter (DAT, Ki = 1 nM, IC50 = 40 and 51nM) with >100 fold selectivity over noradrenalin and 5-HT re-uptake trans

A potent inhibitor of dopamine active transporter (DAT, Ki = 1 nM, IC50 = 40 and 51nM) with >100 fold selectivity over noradrenalin and 5-HT re-uptake transporter. Shown to produce behavioral profile in mice for bipolar mania disease model and ADHD disease models.

Specifications

Content And Storage Protect from light
Form Solid
Quantity 10 mg
Color White
Description 98% by HPLC
Target Proteins DAT
Molecular Formula C28H32F2N2O·2HCl
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.