Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (±)-PD 128907 Hydrochloride

Catalog No. 5058970001 Shop All MilliporeSigma Products
Click to view available options
:
5mg

Selective dopamine D3 receptor agonist (Ki

Selective dopamine D3 receptor agonist (Ki = 2.3nM) with an 18-fold selectivity over long dopamine receptors (D2L) and exhibited only weak affinity for D4.2 receptors (Ki = 169nM). Commonly used to reproduce cocaine′s discriminative stimulus effects and also in peripheral systemic studies of hypertension.

Specifications

Content And Storage Protect from light
Form Powder
Quantity 5 mg
Color White
Description 98% by HPLC
Target Proteins D3
Molecular Formula C14H19NO3·HCl
Solubility H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.