Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PD 168077 Maleate

Catalog No. 5058780001 Shop All MilliporeSigma Products
Click to view available options
:
10mg

A selective D4 dopamine receptor agonist (Ki

A selective D4 dopamine receptor agonist (Ki = 8.7 nM, EC50 = 8.3nM). Suitable for central and peripheral delivery. Used in studies of penile erection, learning and memory. Exhibits some binding affinity towards human D2 and D3 Receptors (Ki = 2.8μM).

Specifications

Content And Storage Protect from light
Form Solid
Quantity 10 mg
Color White
Description 98% by HPLC
Target Proteins D4
Molecular Formula C20H22N4O·C4H4O4
Solubility DMSO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.