Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (-)-Quinpirole Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
:
10mg
Description
A potent, selective dopamine D2 receptor agonist with a Ki value of 4.8 nM. Some D3(Ki = 24nM) and D4 (Ki = 30nM) receptor agonistic action has also been reported. In rat striatum studies in vitro , application of quinpriole inhibited single pulse dopamine overflow in a concentration-dependent manner (IC50 = 32.5nM).
Specifications
Specifications
Form | Powder |
Content And Storage | Protect from light |
Quantity | 10 mg |
Color | White |
Description | 98% by HPLC |
Target Proteins | D2 |
Molecular Formula | C13H21N3·HCl |
Solubility | H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction